Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB5230

Sigma-Aldrich

Anti-Calcium Channel Antibody, Voltage Gated β3

Chemicon®, from rabbit

Synonym(s):

CAB3, Dihydropyridine-sensitive L-type, Calcium Channel beta-3 Subunit, Voltage-dependent Calcium Channel beta-3 Subunit

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$910.00

$910.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
200 μL
$910.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$910.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CACNB3(784)

Specificity

Recognizes the full length Ca beta3 protein, not with other known proteins tested so far.

Immunogen

A purified peptide corresponding to amino acid residues 463-477 of rabbit Ca beta3 (Accession number P54286).
Epitope: Voltage Gated beta3

Application

Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters

Signaling Neuroscience
This Anti-Calcium Channel Antibody, Voltage Gated β3 is validated for use in WB for the detection of Calcium Channel.
Western blot: 1:200-1:300 using ECL on rat brain membranes.

Dilutions should be made using a carrier protein such as BSA (1-3%)

Optimal working dilutions must be determined by the end user.

Physical form

Format: Purified
Purified immunoglobulin. Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, 5% sucrose as a stabilizer and 0.025% sodium azide as a preservative. Reconstitute with 200 μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 xg for 5 min).

Storage and Stability

Maintain lyophilized material at -20°C for up to 12 months. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
CONTROL ANTIGEN: Included free of charge with the antibody is 40 μg of control antigen (lyophilized powder). The stock solution of the antigen can be made up using 100 μL of sterile deionized water. For negative control, preincubate 1 μg of purified peptide with 1 μg of antibody for one hour at room temperature. Optimal concentrations must be determined by the end user.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura A Bell et al.
Journal of neuroinflammation, 17(1), 369-369 (2020-12-05)
Neuron-glial antigen 2 (NG2) cells are a glial cell type tiled throughout the gray and white matter of the central nervous system (CNS). NG2 cells are known for their ability to differentiate into oligodendrocytes and are commonly referred to as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service