Skip to Content
MilliporeSigma
All Photos(1)

Documents

9295-OP

Millipore

TRIS Solution

OmniPur® Grade, 1.0M, pH 9.0

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161700

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

color

clear colorless

shipped in

ambient

General description



Molecular biology grade.

Specifications:
DNase: None detected
Concentration (molarity): 0.9-1.1 M
pH of solution at 25°C: 8.9-9.1
Protease: None detected
RNase: None detected
Sterility: To pass test





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service