Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.51218

Sigma-Aldrich

MPEG-20kDA pNPC

Novabiochem®

Synonym(s):

MPEG-20kDA pNPC, ω-Methoxy-α-propionic acid polyethylene glycol 20 kDa p-nitrophenyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3O(C2H4O)nC3H5O
UNSPSC Code:
51171641

Quality Level

form

powder

manufacturer/tradename

Novabiochem®

storage temp.

15-25°C

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service