Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.02680

Sigma-Aldrich

3-Chloropropionyl chloride

for synthesis

Synonym(s):

3-Chloropropionyl chloride, 3-Chloropropionic acid chloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
126.97
MDL number:
UNSPSC Code:
12352106
EC Index Number:
210-890-4

vapor pressure

10 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

460 °C

potency

1130 mg/kg LD50, oral (Rat)

expl. lim.

8.8-20.2 % (v/v)

pH

<7 ( in H2O)

mp

-32 °C

transition temp

flash point 49 °C

density

1.32 g/cm3 at 20 °C

InChI

1S/C3H4Cl2O/c4-2-1-3(5)6/h1-2H2

InChI key

INUNLMUAPJVRME-UHFFFAOYSA-N

General description

for synthesis

Application

3-Chloropropionyl chloride for synthesis. CAS No. 625-36-5, EC Number 210-890-4.

Analysis Note

Assay (GC, area%)≥ 96.0 % (a/a)Density (d 20 °C/ 4 °C)1.318 - 1.323Identity (IR)passes test

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

158.9 °F - closed cup

Flash Point(C)

70.5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service