Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.01548

Sigma-Aldrich

2-Bromobutane

for synthesis

Synonym(s):

2-Bromobutane, sec-Butyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H9Br
CAS Number:
Molecular Weight:
137.02
MDL number:
UNSPSC Code:
12352101
EC Index Number:
201-140-7
Pricing and availability is not currently available.

vapor pressure

70 hPa ( 20 °C)

Assay

≥98.0% (GC)

autoignition temp.

265 °C

expl. lim.

2.6-6.6 % (v/v)

bp

90-92 °C/1013 hPa

mp

-112 °C

transition temp

flash point 21 °C

solubility

<1 g/L

density

1.26 g/cm3 at 20 °C

storage temp.

2-8°C

InChI

1S/C4H9Br/c1-3-4(2)5/h4H,3H2,1-2H3

InChI key

UPSXAPQYNGXVBF-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.257 - 1.261
Identity (IR): passes test

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

69.8 °F

Flash Point(C)

21 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service