Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

524718

Sigma-Aldrich

Phosphodiesterase Inhibitor Set I

The Phosphodiesterase Inhibitor Set I controls the activity of Phosphodiesterase. This small molecule/inhibitor is primarily used for Cell Signaling applications.

Synonym(s):

PDE Inhibitor Set

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

ambient

storage temp.

2-8°C

General description

A convenient set of inhibitors that are selective for PDE types I, III, IV, and V. Contains 10 mg of 8-Methoxymethyl-3-isobutyl-1-methylxanthine (Cat. No. 454202), a Ca2+/CaM-dependent PDE (PDE I) inhibitor; 1 mg of 4-{[3′,4′-(Methylenedioxy)benzyl]amino}-6-methoxyquinazoline (Cat. No. 475250), a cGMP-specific PDE (PDE V) inhibitor; 5 mg of Rolipram (Cat. No. 557330), a cAMP-specific PDE (PDE IV) inhibitor; and 10 mg of Trequinsin, Hydrochloride (Cat. No. 382425), a cGMP-inhibited PDE (PDE III) inhibitor. Supplied with data sheet.
Phosphodiesterases (PDEs) play an important role in the regulation of cAMP and cGMP levels in the cell. At least five PDE isozymes have been characterized that are differentially expressed and regulated in different cell types: Ca2+-calmodulin-dependent (PDE I); cGMP-stimulated (PDE II); cGMP-inhibited (PDE III); cAMP-specific (PDE IV); cGMP-specific (PDE V). These isozymes exhibit significant homology in their catalytic domain. The cyclic nucleotides are important regulators of many different cellular processes. They are involved in inflammation, asthma, and cardiovascular complications. Hence, specific inhibitors of the PDE isozymes have a potential for use as therapeutic agents. The Phosphodiesterase Inhibitor Set I contains the following inhibitors that are selective for PDE types l, III, IV, and V:



• 8-Methoxymethyl-3-isobutyl-1-methylxanthine(8-Methoxymethyl-IBMX) (Cat. No. 454202), 10 mg

• 4-{[3′,4′-(Methylenedioxy) benzyl]amino}-6-methoxyquinazoline (Cat. No. 475250), 1 mg

• Rolipram (Cat. No. 557330), 5 mg

• Trequinsin, Hydrochloride (Cat. No. 382425), 10 mg

Biochem/physiol Actions

Primary Target
PDE I, PDE III, PDE IV, PDE V

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Reconstitution

Following reconstitution of each inhibitor, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

See properties table for specific solvent and recommended stock concentration.
Takase, Y., et al. 1994. J. Med. Chem.37, 2106.
Underwood, D.C., et al. 1994. J. Pharmacol. Exp. Ther.270, 250.
Wells, J.N. and Miller, J.R. 1988. Methods Enzymol.159, 489.
Ruppert, D. and Weithmann, K.U. 1982. Life Sciences31, 2037.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of serotonin-5-HTR1E signaling pathways and its role in cell survival.
Sharma, et al.
Faseb Journal, 37, e22925-e22925 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service