Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

475980

Sigma-Aldrich

Myoseverin - CAS 267402-71-1 - Calbiochem

Synonym(s):

Myoseverin - CAS 267402-71-1 - Calbiochem, 9-Isopropyl-N²,N⁶- bis-(4-methoxybenzyl)-9H-purine-2,6-diamine, 9-Isopropyl-N²,N⁶-bis-(4-methoxybenzyl)-9H-purine-2,6-diamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H28N6O2
CAS Number:
Molecular Weight:
432.52
UNSPSC Code:
12352200

Assay

≥95% (HPLC)

Quality Level

form

gum (Viscous)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A microtubule-binding molecule that induces reversible fission of multinucleated myotubes into mononucleated fragments. Affects the expression of various growth factors. Induces changes in the expression of genes implicated in extracellular matrix remodeling, stress response, and immunomodulation. May play a role in wound healing and tissue regeneration.
A microtubule-binding molecule that induces reversible fission of multinucleated myotubules into mononucleated fragments. Induces changes in the expression levels of genes implicated in extracellular matrix remodeling, inflammation, coagulation, signal transduction, and cell communication such as those seen in the wounding response of fibroblasts.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Microtubule-binding molecule
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Rosania, G.R., et al. 2000. Nat. Biotechnol.18, 304.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service