Skip to Content
MilliporeSigma
All Photos(2)

Documents

347423

Sigma-Aldrich

β-Glucuronidase Inhibitor

The β-Glucuronidase Inhibitor controls the biological activity of β-Glucuronidase.

Synonym(s):

β-Glucuronidase Inhibitor, 1-((6,8-Dimethyl-2-oxo-1,2-dihydroquinolin-3-yl)methyl)-3-(4-ethoxyphenyl)-1-(2-hydroxyethyl)thiourea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H27N3O3S
CAS Number:
Molecular Weight:
425.54
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.28

Quality Level

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

O=C(N1)C(CN(C(NC2=CC=C(C=C2)OCC)=S)CCO)=CC3=C1C(C)=CC(C)=C3

General description

An orally bioavailable trisubstituted thiourea compound that acts as a potent, uncompetitive and reversible inhibitor of bacterial β-glucuronidase activity (IC50 = 283 nM; kcat/Km = 0.0987 s-1µM-1; Ki = 164 nM in vitro assays against E coli β-glucuronidase; EC50 = 17.7 nM in β-glucuronidase expressing HB101 cells) with excellent selectivity over mammalian β-glucuronidases. Shown to directly target the 17-residue (360-376) loop structure that protects the active site of E coli β-glucuronidase and alter its conformation. Displays negligible cytotoxicity toward both bacterial and mammalian epithelial cells (>>100 µM for HCT116, Caco-2 and CMT93 colon cancer cells) and offers protection against CPT-11 (50 mg/kg, i.p., s.i.d. for 9 days)-induced toxicity in mice (10 µg, p.o., b.i.d).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Wallace, B.D., et al. 2010. Science330, 831.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service