Skip to Content
MilliporeSigma
All Photos(2)

Documents

345815

Millipore

Gentamycin Sulfate, Sterile-Filtered Aqueous Solution, Cell Culture Tested

Gentamicin sulfate is a broad-spectrum cell culture antibiotic that is non-toxic to viruses and mammalian cells at anti-bacterial and anti-mycoplasmal concentrations.

Synonym(s):

Gentamycin Sulfate, Sterile-Filtered Aqueous Solution, Cell Culture Tested, NMD Inhibitor II

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C21H43N5O7.C20H41N5O7.C19H39N5O7.H2O4S/c1-9(25-3)13-6-5-10(22)19(31-13)32-16-11(23)7-12(24)17(14(16)27)33-20-15(28)18(26-4)21(2,29)8-30-20;1-8(21)12-5-4-9(22)18(30-12)31-15-10(23)6-11(24)16(13(15)26)32-19-14(27)17(25-3)20(2,28)7-29-19;1-19(27)7-28-18(13(26)16(19)24-2)31-15-11(23)5-10(22)14(12(15)25)30-17-9(21)4-3-8(6-20)29-17;1-5(2,3)4/h9-20,25-29H,5-8,22-24H2,1-4H3;8-19,25-28H,4-7,21-24H2,1-3H3;8-18,24-27H,3-7,20-23H2,1-2H3;(H2,1,2,3,4)/t9-,10-,11+,12-,13+,14+,15-,16-,17+,18-,19-,20-,21+;8-,9-,10+,11-,12+,13+,14-,15-,16+,17-,18-,19-,20+;8-,9+,10-,11+,12-,13+,14+,15-,16+,17+,18+,19-;/m110./s1

InChI key

RDEIXVOBVLKYNT-HDZPSJEVSA-N

General description

Broad-spectrum cell culture antibiotic that is non-toxic to viruses and mammalian cells at anti-bacterial and anti-mycoplasmal concentrations. Its extended stability and the slow development of bacterial resistance allow long-termvirus and tissue culture studies. Composed of three closely related compounds: gentamycin C1, C1a, and C2.
Broad-spectrum, cell culture antibiotic that is non-toxic to viruses and mammalian cells at anti-bacterial and anti-mycoplasmal concentrations. Its extended stability and the slow development of bacterial resistance allow long-term

virus and tissue culture studies. Composed of three closely related compounds: gentamycin C1, C1a, and C2.

Warning

Toxicity: Harmful (C)

Physical form

Supplied as 50 mg/ml in H₂O.

Reconstitution

Material is autoclavable. Stable over a wide pH range at 37°C for up to 15 days.

Other Notes

Montenez, J.P., et al. 1994. Toxicol. Lett.73, 201.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Antibiotic kill curve is a dose response experiment in which mammalian cells are subjected to increasing amounts of selection antibiotic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service