Skip to Content
MilliporeSigma
All Photos(1)

Documents

300270

Sigma-Aldrich

PARP Inhibitor III, DPQ

The PARP Inhibitor III, DPQ, also referenced under CAS 129075-73-6, controls the biological activity of PARP. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

PARP Inhibitor III, DPQ, 3,4-Dihydro-5[4-(1-piperindinyl)butoxy]-1(2H)-isoquinoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H26N2O2
CAS Number:
Molecular Weight:
302.41
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥90% (HPLC)

form

solid

potency

40 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C18H26N2O2/c21-18-16-7-6-8-17(15(16)9-10-19-18)22-14-5-4-13-20-11-2-1-3-12-20/h6-8H,1-5,9-14H2,(H,19,21)

InChI key

RVOUDNBEIXGHJY-UHFFFAOYSA-N

General description

A potent and selective inhibitor of poly(ADP-ribose) polymerase (PARP; IC50 = 40 nM). Significantly reduces infarcet volume in rat focual cerebrale ischemia model.
A very potent and selective poly(ADP-ribose)polymerase (PARP) inhibitor (IC50 = 40 nM). Significantly reduces infarcet volume in rat focual cerebrale ischemia model.

Biochem/physiol Actions

Cell permeable: no
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Harmful (C)

Other Notes

Eliasson, M.J., et al. 1997. Nat. Med. 3, 1089.
Takahashi, K., et al. 1997. J. Cereb. Blood Flow Metabl. 17, 1137.
Suto, M.J., et al. 1991. Anticancer Drug Des. 6, 107.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service