Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

17-651

Sigma-Aldrich

ChIPAb+ Monomethyl-Histone H4 (Lys20) - ChIP Validated Antibody and Primer Set

from rabbit, purified by affinity chromatography

Synonym(s):

H4K20me1, Histone H4 (mono methyl K20), H4 histone family, member A, histone 1, H4a, histone cluster 1, H4a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.52

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, vertebrates, plant

manufacturer/tradename

ChIPAb+
Upstate®

technique(s)

ChIP: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

General description

All ChIPAb+ antibodies are individually validated for chromatin precipitation, every lot, every time. Each ChIPAb+ antibody set includes control primers (tested every lot by qPCR) to biologically validate your IP results in a locus-specific context. The qPCR protocol and primer sequences are provided, allowing researchers to validate ChIP protocols when using our antibody in their chromatin context. Each set also includes a negative control antibody to ensure specificity of the ChIP reaction.
The ChIPAb+ Monomethyl-Histone H4 (Lys20) set includes the Anti-monomethyl-Histone H4 (Lys20) antibody, a negative control antibody (purified Rabbit IgG), and qPCR primers, which amplify a 212 base pair PCR product within the human GAPDH coding region. The monomethyl histone H4 (Lys20) and negative control antibodies are supplied in a scalable "per ChIP" reaction size and can be used to functionally validate the precipitation of monomethyl-histone H4 (Lys20) associated chromatin.

Specificity

Not tested in other species. The immunogen sequence is identical in a wide range of animal and plant species, so broad cross-reactivity is expected.
Recognizes histone H4, Mr ~11 kDa.

Immunogen

Epitope: a.a. 15-24
The immunogen was a synthetic peptide corresponding to amino acids 15-24 of histone H4, monomethylated on Lys20.

Application

Chromatin Immunoprecipitation:
Sonicated chromatin prepared from HeLa cells (2 X 106 cell equivalents per IP) was subjected to chromatin immunoprecipitation using 4 μg of either a negative control antibody or Anti-Monomethyl-Histone H4
(Lys20) antibody and the Magna ChIP A Kit (Cat. #17-610). Successful immunoprecipitation of monomethyl-histone H4 (Lys20)-associated DNA fragments was verified by qPCR using GAPDH coding region ChIP Primers versus Control Primers corresponding to the GAPDH promoter (Please see figures). Data is presented as percent input of each IP sample relative to input chromatin, with immunoprecipitated DNA from negative control antibody shown as (-) and monomethyl-histone H4 (Lys20) shown as (+).
Please refer to the EZ-Magna ChIP A (Cat. # 17-408) or EZ-ChIP (Cat. # 17-371) protocol for experimental details.

Western Blot Analysis:
0.1 μg/mL of proteins were visualized using a goat anti-rabbit secondary antibody conjugated to HRP and a chemiluminescence detection system (Please see figures)

Peptide Blocking Assay:
40 μg of histone H4 peptide containing monomethyl Lys20 abolished detection of histone H4 by anti-monomethyl-Histone H4 (Lys20) in immunoblot analysis of acid extracts of HeLa cells (Please see figures).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Histones
This ChIPAb+ Monomethyl-Histone H4 (Lys20) -ChIP Validated Antibody & Primer Set conveniently includes the antibody & the specific control PCR primers.

Packaging

25 assays per set. Recommended use: ~4 μg antibody per chromatin immunoprecipitation (dependent upon biological context).

Quality

Chromatin Immunoprecipitation:
Sonicated chromatin prepared from HeLa cells (1 X 106 cell equivalents per IP) were subjected to chromatin immunoprecipitation using 4 μg of either a negative control antibody or Anti-monomethyl-Histone H4 (Lys20) antibody and the Magna ChIP A Kit (Cat. # 17-610). Successful immuno-precipitation of monomethyl-histone H4 (Lys20)-associated DNA fragments was verified by qPCR using ChIP Primers GAPDH Coding region (Please see figures).
Please refer to the EZ-Magna ChIP A (Cat. # 17-408) or EZ-ChIP (Cat. # 17-371) protocol for experimental details.

Target description

~11 kDa

Physical form

Anti-monomethyl-Histone H4 (Lys20) (rabbit polyclonal IgG). One vial containing 100 μg of affinity purified antibody in 100 μL 0.1M Tris-Glycine (pH 7.4), 15 mM NaCl, and 0.05% NaN3. Store at -20°C.

Normal Rabbit IgG. One vial containing 125 μg of purified rabbit IgG in 125 μL storage buffer. Store at -20°C.

ChIP Primers GAPDH Coding Region. One vial containing 75 μL of 5 μM of each primer specific for human GAPDH coding region.Store at -20°C.
FOR: GGC TCC CAC CTT TCT CAT CC
REV: GGC CAT CCA CAG TCT TCT GG

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Includes negative control rabbit IgG antibody and control primers specific for human GAPDH coding region.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chungyu Chang et al.
Current protocols in microbiology, 58(1), e111-e111 (2020-09-01)
This article describes several established approaches for genetic manipulation of Corynebacterium diphtheriae, the causative agent of diphtheria that is known to have provided key evidence for Koch's postulates on the germ theory. First, it includes a detailed gene deletion method
Hua-Rong Zhou et al.
Oncology reports, 37(5), 2663-2671 (2017-04-26)
The present study was designed to investigate the relationship among epigenetic changes in Wnt antagonists, histone H4K20me1 and the expression of tumor-suppressor genes in acute leukemia (AL) to better understand the pathogenesis of leukemia. Quantitative reverse transcription polymerase chain reaction
Karin Meier et al.
PLoS genetics, 8(5), e1002676-e1002676 (2012-05-10)
Mutations in the l(3)mbt tumour suppressor result in overproliferation of Drosophila larval brains. Recently, the derepression of different gene classes in l(3)mbt mutants was shown to be causal for transformation. However, the molecular mechanisms of dL(3)mbt-mediated gene repression are not
Xavier Brenachot et al.
Molecular metabolism, 3(6), 619-629 (2014-08-28)
Overfeeding causes rapid synaptic remodeling in hypothalamus feeding circuits. Polysialylation of cell surface molecules is a key step in this neuronal rewiring and allows normalization of food intake. Here we examined the role of hypothalamic polysialylation in the long-term maintenance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service