Skip to Content
MilliporeSigma
All Photos(1)

Documents

118490

Sigma-Aldrich

ATIC Dimerization Inhibitor

Synonym(s):

ATIC Dimerization Inhibitor, AMPK Signaling Activator XI, Ac-Arg-Ph(4-NO2)-NEt2, ATIC Inhibitor, AICAR Tfase Inhibitor, AICAR transformylase Inhibitor, AICAR Tranformylase/Inosine Monophosphate Cyclohydrolase Dimerization Inhibitor, Ac-Arg-Ph(4-NO2)-NEt2, ATIC Inhibitor, AICAR Tfase Inhibitor, AICAR transformylase Inhibitor, AICAR Tranformylase/Inosine Monophosphate Cyclohydrolase Dimerization Inhibitor, AMPK Signaling Activator XI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H33N7O5
CAS Number:
Molecular Weight:
463.53
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥97% (HPLC)

Quality Level

form

solid

potency

691 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: soluble

shipped in

wet ice

storage temp.

2-8°C

General description

A cell-permeable dipeptide that reversibly blocks the homodimerization of aminoimidazole carboxamide ribonucleotide transformylase/inosine nomophosphate cyclohydrolase (ATIC) and inhibits its activity (Ki = 691 nM, Kd = 240 nM). AITC is a bifunctional enzyme that catalyzes the last two steps of de novo purine biosynthesis. Increases the intracellular levels of AICAR leading to the activation of AMP-activated protein kinase (AMPK). Shown to reduce cell viability in a dose-dependent manner. Exhibits a longer half-life and sustained AMPK activation (~ 24 hours) compared to AICA-riboside (Cat. No. 123040).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
ATIC
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

Ac-Arg-Ph(4-NO2)-NEt2

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Spurr, I.B., et al. 2012. ChemBioChem13, 1628.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service