Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.14556

Supelco

Nitrate Test, photometric

0.10-3.00 mg/L (NO3-N), 0.4-13.3 mg/L (NO3-)

Sign Into View Organizational & Contract Pricing

Select a Size

25 TESTS
$295.00

$295.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
25 TESTS
$295.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

$295.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Product Name

Nitrate Cell Test in seawater, 0.10-3.00 mg/L (NO3-N), 0.4-13.3 mg/L (NO3-)

specific analyte(s)

nitrate

Quality Level

measuring range

0.10-3.00 mg/L (NO3-N)
0.4-13.3 mg/L (NO3-)

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

General description

Method: photometric 0.10 - 3.00 mg/l NO₃-N 0.4 - 13.3 mg/l NO₃⁻ Spectroquant®
This Spectroquant® Nitrate Cell Test is specially designed for the analysis of low levels of nitrate in seawaters or other water samples with high loads of chloride. Nitrate is an essential nutrient required for the growth of animals and plants. However, elevated levels of nitrate can be a health risk for animals and humans, in particular for pregnant women and infants.
The Spectroquant® Cell Tests come with prefilled 16 mm round cells and all the required reagents to perform the analysis according to the instruction leaflet provided.
All Spectroquant® Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1 - Skin Sens. 1 - STOT SE 2 Oral

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service