Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.14237

Supelco

Spare filters for EXtrelut® NT3 prepacked glass columns

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

technique(s)

supported liquid extraction (SLE): suitable

Quality Level

storage temp.

no temp limit

General description

Extrelut® NT simplifies liquid-liquid extraction in its most effective form by using a chemically inert, wide-pore and highly pure diatomaceous earth based solid phase. Depending on the sample volume three types of columns are available: Extrelut NT1 can take up a maximum of one ml, Extrelut NT3 three ml and Extrelut NT20 up to twenty ml of sample.

Analysis Note

visual control: passes test

Legal Information

Extrelut is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Simplify your liquid-liquid extractions with our emulsion-free supported-liquid extraction (SLE) single step solution.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service