Skip to Content
MilliporeSigma
All Photos(2)

Documents

1.09687

Supelco

n-Hexane

reference substance for gas chromatography

Synonym(s):

Hexane, n-Hexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH3
CAS Number:
Molecular Weight:
86.18
Beilstein:
1730733
MDL number:
UNSPSC Code:
12352001
EC Index Number:
203-777-6

vapor density

~3 (vs air)

Quality Level

vapor pressure

256 mmHg ( 37.7 °C)
5.2 psi ( 37.7 °C)
~132 mmHg ( 20 °C)

Assay

≥99.7% (GC)

form

liquid

autoignition temp.

453 °F

potency

25000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

expl. lim.

7.7 %

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.375 (lit.)

kinematic viscosity

0.50 cSt(20 °C)

bp

69 °C (lit.)

mp

−95 °C (lit.)

transition temp

flash point -22 °C

solubility

0.0095 g/L

density

0.659 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

CCCCCC

InChI

1S/C6H14/c1-3-5-6-4-2/h3-6H2,1-2H3

InChI key

VLKZOEOYAKHREP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Linkage

Replaces: 9687-2

Analysis Note

Assay (GC): ≥ 99.7 %
Identity (IR): conforms

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-7.6 °F

Flash Point(C)

-22 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service