Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.09678

Supelco

Dimethyl sulfoxide

for gas chromatography

Synonym(s):

Methanesulfinylmethane, Methyl sulfoxide, Dimethyl(oxido)sulfur, DMSO, DMSO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2SO
CAS Number:
Molecular Weight:
78.13
Beilstein:
506008
MDL number:
UNSPSC Code:
12352119
EC Index Number:
200-664-3

vapor density

2.7 (vs air)

Quality Level

vapor pressure

0.42 mmHg ( 20 °C)

Assay

≥99.7% (GC)

form

liquid

autoignition temp.

573 °F

potency

28300 mg/kg LD50, oral (Rat)
40000 mg/kg LD50, skin (Rat)

expl. lim.

42 %, 63 °F

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.479 (lit.)

kinematic viscosity

2.14 cSt

bp

189 °C (lit.)

mp

16-19 °C (lit.)

transition temp

flash point 87 °C

density

1.10 g/mL (lit.)

storage temp.

2-30°C

SMILES string

CS(C)=O

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3

InChI key

IAZDPXIOMUYVGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application



  • In Vitro Inhibition of Colorectal Cancer Gene Targets by Withania somnifera L. Methanolic Extracts: A Focus on Specific Genome Regulation. Research indicates the use of DMSO as a solvent in extracting phytochemicals for colorectal cancer research, emphasizing its importance in pharmaceutical studies and drug discovery processes (Macharia JM, Pande DO, Zand A, et al. 2024).

  • Nobiletin as a novel agent to enhance porcine in vitro embryo development and quality. The study uses DMSO as a solvent to study the effects of Nobiletin, highlighting its utility in developmental biology and reproductive technology (Cajas YN, Cañón-Beltrán K, Mazzarella R, et al. 2024).

  • Improving the Monophenolic Yield of Lignin Depolymerization in Dualistic Aprotic Solvent System by Organic Solvent Fractionation. This article explores the solvent properties of DMSO in enhancing lignin depolymerization, underlining its critical role in green chemistry and sustainability studies (Xu Y, Dai Q, Zhang D, et al. 2024).

  • Inter-coat protein loading of active ingredients into Tobacco mild green mosaic virus through partial dissociation and reassembly of the virion. This research utilizes DMSO for the manipulation of viral vectors in genetic engineering, illustrating its versatility in molecular biology applications (González-Gamboa I, Caparco AA, McCaskill J, et al. 2024).




Caution

Supercools easily and remelts slowly at room temperature. Solidified product can be re-liquified by warming to room temperature without detriment to the product.

Specifications

Saturation Concentration in Air: 8.0 g/m3 (20 °C)

Analysis Note

Assay (GC): ≥ 99.7 %
Identity (IR): conforms
Water: ≤ 0.3 %

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service