Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.00586

Supelco

Ion exchanger Dowex Monosphere 650C (H)

suitable for ion chromatography

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41115700

form

beads

technique(s)

HPLC: suitable
LPLC: suitable
ion chromatography: suitable

matrix

PS-DVB

matrix active group

SCX

particle size

300-1100 μm

separation technique

cation exchange

General description

(strongly acidic cation exchanger, H⁺ form)

Legal Information

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service