Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

03-309

Sigma-Aldrich

Magna ChIRP® TERC lncRNA Probe Set

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

technique(s)

DNA sequencing: suitable
nucleic acid detection: suitable

Quality Level

suitability

suitable for determination of protein concentration

General description

ChIRP (Chromatin Isolation by RNA Purification), allows recovery of specific segments of DNA and proteins using chromatin associated RNA as a target for biotinylated antisense probes. The Magna ChIRP TERC lncRNA Probe Set contains 19 predesigned 20-mer DNA oligonucleotides tiled along and complementary to the sequence of human lncRNA TERC. All probes are biotin-TEG-labeled at the 3-prime end. The oligonucleotides are split to two pools (even and odd) and are supplied ready-for-use by the ChIRP method. This probe set is fully compatible with the Maga ChIRP kits (cat. # 17-10494 and 17-10495).

Physical form

50 µM biotin-TEG-labeled 20-mer DNA oligonucleotides in containing 10 mM Tris-HCl, 1mM EDTA, pH 8.0

Storage and Stability

Upon receipt store at -20°C. Avoid multiple freeze thaws. With proper storage and handling this product is stable for 6 months from the date of receipt.

Legal Information

MAGNA CHIRP is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service