Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAT370174947

Matrix Scientific

1-[(4-Chlorophenyl)methyl]-5,5-dimethylimidazolidin-2-one

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
238.72
MDL number:

mp

122-126

Iupac Name

1-(4-chlorobenzyl)-5,5-dimethyl-2-imidazolidinone

Inchi Code

1S/C12H15ClN2O/c1-12(2)8-14-11(16)15(12)7-9-3-5-10(13)6-4-9/h3-6H,7-8H2,1-2H3,(H,14,16)

InChI key

XUDCGBBALDNPIO-UHFFFAOYSA-N

Purity

95%

Country of Origin

US

Product Link

https://www.matrixscientific.com/076621.html

Melting Point

122-126

Pictograms

MSDS

https://www.matrixscientific.com/076621.html


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service