Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAN412118847

Manchester Organics

Propyl methanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
138.19
MDL number:

Iupac Name

propyl methanesulfonate

Inchi Code

1S/C4H10O3S/c1-3-4-7-8(2,5)6/h3-4H2,1-2H3

InChI key

DKORSYDQYFVQNS-UHFFFAOYSA-N

Country of Origin

GB

Product Link

https://www.manchesterorganics.com/S40933

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service