Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

LIF372152484

Life Chemicals Inc.

4-Hydroxy-benzoic acid 2,2,3,3-tetrafluoro-propyl ester

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
252.17
MDL number:

Iupac Name

2,2,3,3-tetrafluoropropyl 4-hydroxybenzoate

Inchi Code

1S/C10H8F4O3/c11-9(12)10(13,14)5-17-8(16)6-1-3-7(15)4-2-6/h1-4,9,15H,5H2

InChI key

YGEVQEXZCKHLCD-UHFFFAOYSA-N

Purity

95%

Country of Origin

UA

Physical Form

solid

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service