Skip to Content
MilliporeSigma

KEYH57C560DF

Key Organics / BIONET

4-chloro-2-[(Z)-(2-chlorophenyl)(hydroxyimino)methyl]aniline

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
281.14
MDL number:

Iupac Name

4-chloro-2-[(2-chlorophenyl)(nitroso)methyl]aniline

Inchi Code

1S/C13H10Cl2N2O/c14-8-5-6-12(16)10(7-8)13(17-18)9-3-1-2-4-11(9)15/h1-7,13H,16H2

InChI key

HDYRMVOBYBUCED-UHFFFAOYSA-N

Purity

95%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/SS-5272

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary statements

MSDS

https://www.keyorganics.net/sds/SS-5272


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service