Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

KEY771306146

Key Organics / BIONET

imino(methyl)(4-methylphenyl)-lambda6-sulfanone

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
169.25
MDL number:

Iupac Name

1-methyl-4-(methylsulfonimidoyl)benzene

Inchi Code

1S/C8H11NOS/c1-7-3-5-8(6-4-7)11(2,9)10/h3-6H,2H2,1H3,(H2,9,10)

InChI key

PXZLDSLFQYGNJM-UHFFFAOYSA-N

Purity

95%

Country of Origin

GB

Physical Form

Liquid,Solid

Product Link

https://www.keyorganics.net/sku/SS-4987

Pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.keyorganics.net/sds/SS-4987


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service