Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

KEY514453019

Key Organics / BIONET

3,5-dibromo-1H-indazole

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
275.93
MDL number:

Iupac Name

3,5-dibromo-1H-indazole

Inchi Code

1S/C7H4Br2N2/c8-4-1-2-6-5(3-4)7(9)11-10-6/h1-3H,(H,10,11)

InChI key

YPNFVZZHYXRCKH-UHFFFAOYSA-N

Purity

95%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/GS-4407

Pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.keyorganics.net/sds/GS-4407


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service