Skip to Content
MilliporeSigma
All Photos(1)

Documents

KEY370561183

Key Organics / BIONET

1-carbamoylcyclobutane-1-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
143.14
MDL number:

mp

174 - 176

Iupac Name

1-(aminocarbonyl)cyclobutanecarboxylic acid

Inchi Code

1S/C6H9NO3/c7-4(8)6(5(9)10)2-1-3-6/h1-3H2,(H2,7,8)(H,9,10)

InChI key

IMVNGQDQHJICRV-UHFFFAOYSA-N

Purity

95%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/FG-0705

Melting Point

174 - 176

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

MSDS

https://www.keyorganics.net/sds/FG-0705


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service