Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

KEY320856985

Key Organics / BIONET

2,2-bis(2-chloro-5-nitrophenyl)acetonitrile hydrate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
370.15
MDL number:

mp

209 - 211

Iupac Name

bis(2-chloro-5-nitrophenyl)acetonitrile hydrate

Inchi Code

1S/C14H7Cl2N3O4.H2O/c15-13-3-1-8(18(20)21)5-10(13)12(7-17)11-6-9(19(22)23)2-4-14(11)16;/h1-6,12H;1H2

InChI key

BOEONDOHCYIJPA-UHFFFAOYSA-N

Purity

95%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/BF-0700

Melting Point

209 - 211

Pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.keyorganics.net/sds/BF-0700


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service