Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

KEY195583169

Key Organics / BIONET

(6-bromo-1H-indol-2-yl)methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
226.07
MDL number:
Pricing and availability is not currently available. Contact Technical Service

mp

111 - 113

Iupac Name

(6-bromo-1H-indol-2-yl)methanol

Inchi Code

1S/C9H8BrNO/c10-7-2-1-6-3-8(5-12)11-9(6)4-7/h1-4,11-12H,5H2

InChI key

CNSVVCNALIWZAK-UHFFFAOYSA-N

storage temp.

Store under inert gas at 4C

Purity

95%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/ED-0743

Melting Point

111 - 113

Pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.keyorganics.net/sds/ED-0743


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service