Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

JWPH98D3FCE8

J & W Pharmlab LLC

Potassium trifluoro[3-(methoxycarbonyl)bicyclo[1.1.1]pent-1-yl]-borate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
232.05
MDL number:

Iupac Name

potassium trifluoro(3-(methoxycarbonyl)bicyclo[1.1.1]pentan-1-yl)borate

Inchi Code

1S/C7H9BF3O2.K/c1-13-5(12)6-2-7(3-6,4-6)8(9,10)11;/h2-4H2,1H3;/q-1;+1

InChI key

ASAXRQFLFVABCA-UHFFFAOYSA-N

Purity

96%

Country of Origin

US

Physical Form

white solid

Product Link

https://www.jwpharmlab.com/home/product/32R0407K

MSDS

https://www.jwpharmlab.com/home/product/32R0407K


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service