Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

JWPH93D8B448

J & W Pharmlab LLC

Pyridin-2-yl-glycine tert-butyl ester hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
244.72
MDL number:

Iupac Name

tert-butyl 2-amino-2-(pyridin-2-yl)acetate hydrochloride

Inchi Code

1S/C11H16N2O2.ClH/c1-11(2,3)15-10(14)9(12)8-6-4-5-7-13-8;/h4-7,9H,12H2,1-3H3;1H

InChI key

DBHHTOORADUNGR-UHFFFAOYSA-N

Purity

97%

Country of Origin

US

Physical Form

Off White Solid

Product Link

https://www.jwpharmlab.com/home/product/15R0144S

MSDS

https://www.jwpharmlab.com/home/product/15R0144S


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service