Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENAH987FB41C

Enamine

10-[(3,4,5-trimethoxyphenyl)methylidene]-9,10-dihydroanthracen-9-one

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
372.42
MDL number:
Pricing and availability is not currently available. Contact Technical Service

mp

176-177

Iupac Name

10-(3,4,5-trimethoxybenzylidene)anthracen-9(10H)-one

Inchi Code

1S/C24H20O4/c1-26-21-13-15(14-22(27-2)24(21)28-3)12-20-16-8-4-6-10-18(16)23(25)19-11-7-5-9-17(19)20/h4-14H,1-3H3

InChI key

JWHMRMLZXWDBCZ-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-100442

Melting Point

176-177

MSDS

https://www.enaminestore.com/catalog/EN300-100442


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service