Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENAH9453D8F9

Enamine

6-chloropyridine-3-sulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
193.61
MDL number:

mp

242-243

Iupac Name

6-chloro-3-pyridinesulfonic acid

Inchi Code

1S/C5H4ClNO3S/c6-5-2-1-4(3-7-5)11(8,9)10/h1-3H,(H,8,9,10)

InChI key

NWKWBQDPHGDVAI-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-6769787

Melting Point

242-243

MSDS

https://www.enaminestore.com/catalog/EN300-6769787


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service