Skip to Content
MilliporeSigma
All Photos(1)

Documents

ENAH65AFCD48

Enamine

pentacyclo[7.3.1.1^{4,12}.0^{2,7}.0^{6,11}]tetradecane-4,9-diamine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
218.34

Iupac Name

(4r,4ar,8r,8ar,10r,11r)-octahydro-2,8,4,6-(epibutane[1,2,3,4]tetrayl)naphthalene-2,6-diamine

Inchi Code

1S/C14H22N2/c15-13-1-7-8-4-14(16)5-9(7)11(3-13)12(6-14)10(8)2-13/h7-12H,1-6,15-16H2

InChI key

OQFGCBHYTPVGEY-UHFFFAOYSA-N

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-7471046

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service