Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENAH6592797F

Enamine

1-methyl-2-sulfanylidene-1,3-diazinane-4,6-dione

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
158.18
MDL number:

mp

197-202

Iupac Name

1-methyl-2-thioxodihydro-4,6(1H,5H)-pyrimidinedione

Inchi Code

1S/C5H6N2O2S/c1-7-4(9)2-3(8)6-5(7)10/h2H2,1H3,(H,6,8,10)

InChI key

DLMPJSWFKZILOI-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-7239991

Melting Point

197-202

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-7239991


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service