Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENAH304B3854

Enamine

4-(piperidin-4-yloxy)benzene-1,3-diol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
209.24

Iupac Name

4-(piperidin-4-yloxy)benzene-1,3-diol

Inchi Code

1S/C11H15NO3/c13-8-1-2-11(10(14)7-8)15-9-3-5-12-6-4-9/h1-2,7,9,12-14H,3-6H2

InChI key

SRGUKJORKSTMFX-UHFFFAOYSA-N

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-1764814

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service