Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENAH30486264

Enamine

[2-(chloromethyl)pentyl]cyclopentane

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
188.74
MDL number:

Iupac Name

[2-(chloromethyl)pentyl]cyclopentane

Inchi Code

1S/C11H21Cl/c1-2-5-11(9-12)8-10-6-3-4-7-10/h10-11H,2-9H2,1H3

InChI key

UMQIFMAGWGZOPC-UHFFFAOYSA-N

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-676705

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service