ENA964468307
Enamine
N-phenylazetidin-3-amine dihydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
N-phenylazetidin-3-amine dihydrochloride
Inchi Code
1S/C9H12N2.2ClH/c1-2-4-8(5-3-1)11-9-6-10-7-9;;/h1-5,9-11H,6-7H2;2*1H
InChI key
FFBCUXLAGFRVAW-UHFFFAOYSA-N
storage temp.
4
Salt Data
Cl
Purity
95
Country of Origin
UA
Shipping Temperature
Normal
Physical Form
powder
Product Link
https://www.enaminestore.com/catalog/EN300-650551
Signal Word
Warning
Hazard Statements
MSDS
https://www.enaminestore.com/catalog/EN300-650551
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service