Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENA946832034

Enamine

tert-butyl N-{[methoxy(methyl)carbamoyl]methyl}-N-methylcarbamate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
232.28
MDL number:

Iupac Name

tert-butyl 2-[methoxy(methyl)amino]-2-oxoethyl(methyl)carbamate

Inchi Code

1S/C10H20N2O4/c1-10(2,3)16-9(14)11(4)7-8(13)12(5)15-6/h7H2,1-6H3

InChI key

AWJCGVOYTACHLS-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.enaminestore.com/catalog/EN300-331635

Pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.enaminestore.com/catalog/EN300-331635


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service