ENA447957007
Enamine
3-amino-1-(2-aminophenyl)propan-1-one dihydrochloride
About This Item
mp
185-188
Iupac Name
3-amino-1-(2-aminophenyl)propan-1-one dihydrochloride
Inchi Code
1S/C9H12N2O.2ClH/c10-6-5-9(12)7-3-1-2-4-8(7)11;;/h1-4H,5-6,10-11H2;2*1H
InChI key
GIUIHRRGPBPUAO-UHFFFAOYSA-N
storage temp.
Salt Data
Purity
Country of Origin
Shipping Temperature
Physical Form
Product Link
Melting Point
Signal Word
Warning
Hazard Statements
MSDS
https://www.enaminestore.com/catalog/EN300-207293
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service