Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENA429358538

Enamine

2,4-dichloro-5-iodobenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
316.91
MDL number:

Iupac Name

2,4-dichloro-5-iodobenzoic acid

Inchi Code

1S/C7H3Cl2IO2/c8-4-2-5(9)6(10)1-3(4)7(11)12/h1-2H,(H,11,12)

InChI key

RVEQDEIBFPXXBJ-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-183313

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-183313


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service