Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENA388191749

Enamine

2-bromo-4H,5H,6H,7H-thieno[2,3-c]pyridine hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
299.03
MDL number:

mp

226-228

Iupac Name

2-bromo-4,5,6,7-tetrahydrothieno[2,3-c]pyridine hydrobromide

Inchi Code

1S/C7H8BrNS.BrH/c8-7-3-5-1-2-9-4-6(5)10-7;/h3,9H,1-2,4H2;1H

InChI key

LYFTUUNJHCCTAO-UHFFFAOYSA-N

storage temp.

4

Salt Data

Br

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-120711

Melting Point

226-228

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-120711


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service