Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENA181911536

Enamine

(5-bromofuran-2-yl)methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
177
MDL number:

mp

43-44

Iupac Name

(5-bromo-2-furyl)methanol

Inchi Code

1S/C5H5BrO2/c6-5-2-1-4(3-7)8-5/h1-2,7H,3H2

InChI key

CFLMVPYEVZXIED-UHFFFAOYSA-N

storage temp.

-70

Purity

95

Country of Origin

UA

Shipping Temperature

Dry Ice

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-37107

Melting Point

43-44

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

MSDS

https://www.enaminestore.com/catalog/EN300-37107


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service