Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

COM448643704

Combi-Blocks, Inc.

Perfluorophenyl 4-fluorobenzoate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
306.16
MDL number:

Iupac Name

2,3,4,5,6-pentafluorophenyl 4-fluorobenzoate

Inchi Code

1S/C13H4F6O2/c14-6-3-1-5(2-4-6)13(20)21-12-10(18)8(16)7(15)9(17)11(12)19/h1-4H

InChI key

AEMGSQVSVDUWQG-UHFFFAOYSA-N

Purity

97%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?QA-9253

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?QA-9253


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service