Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

COM448616379

Combi-Blocks, Inc.

2-Acrylamidophenylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
190.99
MDL number:

Iupac Name

2-(acryloylamino)phenylboronic acid

Inchi Code

1S/C9H10BNO3/c1-2-9(12)11-8-6-4-3-5-7(8)10(13)14/h2-6,13-14H,1H2,(H,11,12)

InChI key

XGDUBFMBDDZYKB-UHFFFAOYSA-N

storage temp.

Refrigerated

Purity

97%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?BB-9121

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?BB-9121


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service