Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CIAH98ABF3AE

ChemScene LLC (Preferred Partner)

2-Chloro-4,5-dimethylaniline

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
155.63
MDL number:

bp

248.7+-35.0 C at 760 mmHg

mp

85.0-86.5 C

Iupac Name

2-chloro-4,5-dimethylaniline

Inchi Code

1S/C8H10ClN/c1-5-3-7(9)8(10)4-6(5)2/h3-4H,10H2,1-2H3

InChI key

CJZAXFZHDSZLRR-UHFFFAOYSA-N

storage temp.

4C, protect from light

Purity

98%

Country of Origin

CN

Physical Form

Solid

Product Link

https://www.chemscene.com/1585-13-3.html

Boiling point

248.7+-35.0 C at 760 mmHg

Melting Point

85.0-86.5 C

Signal Word

Warning

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0061243.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service