Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CIAH987F34D4

ChemScene LLC (Preferred Partner)

Ethyl 5-methylfuran-3-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
154.17
MDL number:
Pricing and availability is not currently available. Contact Technical Service

bp

69-71 C at 6 mmHg

Iupac Name

ethyl 5-methyl-3-furoate

Inchi Code

1S/C8H10O3/c1-3-10-8(9)7-4-6(2)11-5-7/h4-5H,3H2,1-2H3

InChI key

HZVHSZJXVCZRTP-UHFFFAOYSA-N

storage temp.

4C

Purity

98%

Country of Origin

CN

Physical Form

Liquid

Product Link

https://www.chemscene.com/26501-83-7.html

Boiling point

69-71 C at 6 mmHg

Signal Word

Danger

Hazard Statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0158923.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service