Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CH6447917518

Chem Impex International, Inc.

(3-Pyridylmethyl)Sulfonyl Chloride Triflate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
341.72
MDL number:

Iupac Name

trifluoromethanesulfonic acid compound with 3-pyridinylmethanesulfonyl chloride (1:1)

Inchi Code

1S/C6H6ClNO2S.CHF3O3S/c7-11(9,10)5-6-2-1-3-8-4-6;2-1(3,4)8(5,6)7/h1-4H,5H2;(H,5,6,7)

InChI key

FTWJYPAJVLWLAG-UHFFFAOYSA-N

storage temp.

Store at 0-8 C

Purity

95%

Country of Origin

US

Shipping Temperature

RT

Physical Form

Semi solid

Product Link

https://www.chemimpex.com/product/productinfo/3-pyridylmethyl-sulfonyl-chloride-triflate/14047?cid=0

MSDS

https://www.chemimpex.com/product/productinfo/3-pyridylmethyl-sulfonyl-chloride-triflate/14047?cid=0


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service