Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CH6371387496

Chem Impex International, Inc.

1-(2,4-Difluorophenyl)-3-(thiophen-2-yl)-1H-pyrazole-5-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
306.29
MDL number:

mp

225 - 230 C

Iupac Name

1-(2,4-difluorophenyl)-3-(2-thienyl)-1H-pyrazole-5-carboxylic acid

Inchi Code

1S/C14H8F2N2O2S/c15-8-3-4-11(9(16)6-8)18-12(14(19)20)7-10(17-18)13-2-1-5-21-13/h1-7H,(H,19,20)

InChI key

ULPHWQUOXQAERG-UHFFFAOYSA-N

storage temp.

Store at 0 - 8 C

Purity

98%

Country of Origin

US

Shipping Temperature

RT

Physical Form

Off-white powder

Product Link

https://www.chemimpex.com/product/productinfo/1-2-4-difluorophenyl-3-thiophen-2-yl-1-i-h-i-pyrazole-5-carboxylic-acid/16360?cid=0

Melting Point

225 - 230 C

MSDS

https://www.chemimpex.com/product/productinfo/1-2-4-difluorophenyl-3-thiophen-2-yl-1-i-h-i-pyrazole-5-carboxylic-acid/16360?cid=0


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service