Skip to Content

We are planning system maintenance on January 25, 2025, from 1:00 PM to 4:00 PM CST. This will impact both web and offline transactions, including online orders, quotes, price and availability checks, and order status inquiries. We apologize for any inconvenience.

MilliporeSigma
All Photos(1)

Key Documents

C12H94457AD0

Chemcia Scientific, LLC

3-Nitro-1,4-dihydro-pyrrolo[3,2-b]pyridin-5-one

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
179.13

Pricing and availability is not currently available. Contact Technical Service

Iupac Name

3-nitro-1H-pyrrolo[3,2-b]pyridin-5-ol

Inchi Code

1S/C7H5N3O3/c11-6-2-1-4-7(9-6)5(3-8-4)10(12)13/h1-3,8H,(H,9,11)

InChI key

YFXFZKDCDNZHLV-UHFFFAOYSA-N

storage temp.

0-5C

Purity

97%

Country of Origin

US

Shipping Temperature

Ambient

Physical Form

Yellow Solid

Product Link

http://www.chemcia.com/product_fl02.asp?id=13457

MSDS

http://www.chemcia.com/product_fl02.asp?id=13457


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service