Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CH4415172786

ChemBridge Corporation

[2-(2-propyn-1-yloxy)phenyl]amine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
183.64
MDL number:

Iupac Name

2-(prop-2-yn-1-yloxy)aniline hydrochloride

Inchi Code

1S/C9H9NO.ClH/c1-2-7-11-9-6-4-3-5-8(9)10;/h1,3-6H,7,10H2;1H

InChI key

TVSAEWXHJPZLKN-UHFFFAOYSA-N

storage temp.

RT

Salt Data

HCl

Purity

95%

Country of Origin

KZ

Shipping Temperature

RT

Physical Form

solid

Product Link

https://www.hit2lead.com/building-blocks/4088026

MSDS

https://www.hit2lead.com/sds/4088026_sds.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service