Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900129E

Avanti

15(S)-HAEA

Avanti Research - A Croda Brand (900129E), ethanol solution

Synonym(s):

15(S)-hydroxy-N-(2-hydroxyethyl)-5Z,8Z,11Z,13E-eicosatetraenamide

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$251.00
250 μG
$933.00

$251.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
$251.00
250 μG
$933.00

About This Item

Empirical Formula (Hill Notation):
C22H37NO3
CAS Number:
Molecular Weight:
363.53
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$251.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

ethanol solution

packaging

pkg of 1 × 250 μg (900129E-250UG)
pkg of 1 × 50 μg (900129E-50UG)

manufacturer/tradename

Avanti Research - A Croda Brand (900129E)

concentration

250 μg/mL in ethanol (900129E-250UG)
50 μg/mL in ethanol (900129E-50UG)

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCC[C@H](O)/C=C/C=C\C/C=C\C/C=C\CCCC(NCCO)=O

InChI

1S/C22H37NO3/c1-2-3-13-16-21(25)17-14-11-9-7-5-4-6-8-10-12-15-18-22(26)23-19-20-24/h4-5,8-11,14,17,21,24-25H,2-3,6-7,12-13,15-16,18-20H2,1H3,(H,23,26)/b5-4-,10-8-,11-9-,17-14+/t21-/m0/s1

InChI key

XZQKRCUYLKDPEK-BPVVGZHASA-N

General description

15-HAEA is an inhibitor of fatty acid amide hydrolase (FAAH).[1]

Packaging

2 mL Amber Glass Sealed Ampule (900129E-50UG, 900129E-250UG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Methylation and acetylation of 15-hydroxyanandamide modulate its interaction with the endocannabinoid system
Amadio D, et al.
Biochimie, 92(4), 378-387 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service